Key Documents
BR144605
BRAND® BISTABIL® ground glass stopper, conical joint
joint: ST/NS 7/16
Synonym(s):
BRAND® BISTABIL® ground glass stopper conical joint stopper, Conical joint stoppers, Stoppers
Select a Size
1 EA
$11.80
$11.80
Check Cart for Availability
Select a Size
Change View
1 EA
$11.80
About This Item
UNSPSC Code:
41122500
NACRES:
NB.51
$11.80
Check Cart for Availability
Joint
joint: ST/NS7/16
description
hexagonal grip
hollow
packaging
pack of 1 ea
manufacturer/tradename
BRAND 144605
General description
Conical ground joint stopper made from Duran®. Hollow with drip tip and hexagonal grip. Meets DIN 12 252.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Bistabil is a registered trademark of Lenz GmbH
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)