Key Documents
BR136110
BRAND® stirring spatula
polypropylene
Synonym(s):
Stirring rods
Select a Size
10 EA
$41.50
$41.50
Available to ship on14 April 2025Details
Select a Size
Change View
10 EA
$41.50
About This Item
UNSPSC Code:
41122403
NACRES:
NB.43
$41.50
Available to ship on14 April 2025Details
material
polypropylene
packaging
pack of 10 ea
manufacturer/tradename
BRAND 136110
L × D
245 mm × 6 mm
General description
The BRAND® stirring spatula is made from durable polypropylene (PP) and features two differently shaped ends, each approximately 15 mm wide. This versatile design allows for effective mixing, stirring, and scraping of various substances in laboratory applications. The lightweight construction makes it easy to handle, while the chemical resistance of PP ensures durability against a range of reagents.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)