Skip to Content
Merck
All Photos(1)

Key Documents

BR115515

BRAND® test tube rack

PTFE, Holds 10 x 19 mm tubes

Synonym(s):

Accessories, Test Tube Racks, Tubes

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$305.00

$305.00


Check Cart for Availability


Select a Size

Change View
1 EA
$305.00

About This Item

UNSPSC Code:
41122804
NACRES:
NB.35

$305.00


Check Cart for Availability

material

PTFE

feature

Holds 10 x 19 mm tubes

packaging

pack of 1 ea

manufacturer/tradename

BRAND 115515

L × W × H

180 mm × 60 mm × 70 mm

Looking for similar products? Visit Product Comparison Guide

General description

The BRAND® test tube rack is constructed from high-quality PTFE, ensuring excellent chemical resistance for a variety of laboratory applications. This robust rack is designed to withstand operating temperatures ranging from -200 to 250 °C, making it suitable for both extreme cold and high-heat environments. Its durable design provides secure support for test tubes, preventing spills and breakage during experiments. The non-reactive nature of PTFE ensures that samples remain uncontaminated, enhancing the reliability of your laboratory work.

Features and Benefits

  • BRAND® test tube racks are sturdy with excellent mechanical strength.
  • Chemically resistant to most reagents.
  • Suitable for use in temperatures from -200 °C to 250 °C.

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service