Key Documents
BR114720
BRAND® grip stopper for sample and sedimentation tubes
for sample tubes BR114715
Synonym(s):
Accessories, Stoppers, Test tubes, Tubes
Select a Size
1000 EA
$64.10
$64.10
Available to ship on28 April 2025Details
Select a Size
Change View
1000 EA
$64.10
About This Item
UNSPSC Code:
41121800
NACRES:
NB.11
$64.10
Available to ship on28 April 2025Details
material
low-density polyethylene
packaging
pack of 1000 ea
manufacturer/tradename
BRAND 114720
Related Categories
General description
The BRAND® Grip Stopper is specifically designed for use with sample and sedimentation tubes 114715, providing a secure and effective sealing solution. Made from neutral PE-LD (low-density polyethylene), this stopper ensures compatibility with various substances while maintaining a tight seal to prevent contamination. Its ergonomic design allows for easy handling and application, making it ideal for laboratory environments. This grip stopper enhances the functionality of your tubes, ensuring safe storage and transport of samples and liquids while facilitating reliable sedimentation processes.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)