Skip to Content
Merck
All Photos(1)

Key Documents

BMSBT4500E

Benchmark Orbi-Shaker CO2 with remote controller

AC/DC input 230 V, EuroPlug

Synonym(s):

orbital shaker, microplate shaker, carbon dioxide incubator, CO2 incubation, harsh enviroment incubation

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$6,650.00

$6,650.00


Check Cart for Availability


Select a Size

Change View
1 EACH
$6,650.00

About This Item

UNSPSC Code:
41103811
NACRES:
NB.73

$6,650.00


Check Cart for Availability

manufacturer/tradename

Benchmark Scientific BT4500-E

parameter

200-1200 rpm speed

plate W × L

33 cm × 30 cm

Looking for similar products? Visit Product Comparison Guide

General description

OrbiShaker CO2MP is designed for thorough, mixing of microplates in extreme environments, including CO2 incubators. It can withstand temperatures from 0 to 65°C and humidity levels up to 100%.

Application

Accepts 6 microplates, designed for use in CO2 incubators

Legal Information

Orbi-Shaker is a trademark of Benchmark Scientific, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service