BAH420530000
SP Bel-Art® Dry-Keeper Small, Stacking Desiccator Cabinet
polystyrene, volume 0.14 ft3
Select a Size
$464.00
Estimated to ship on26 March 2025
Select a Size
About This Item
$464.00
Estimated to ship on26 March 2025
Recommended Products
material
polystyrene
packaging
pkg of 1 ea
manufacturer/tradename
Bel-Art H42053-0000
volume
0.14 ft3
General description
- Pull down door slides out of the way into a compartment located underneath the cabinet
- Ready to use jar of silica gel is included
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)