Skip to Content
Merck
All Photos(1)

Documents

BAH379220000

SP Bel-Art® Fluorocarbon Plastic Forceps

plastic forceps (fluorocarbon), L 4.5 in. (114 mm)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122800
NACRES:
NB.43

material

plastic forceps (fluorocarbon)

packaging

pkg of 1 ea

manufacturer/tradename

Bel-Art H37922-0000

L

4.5 in. (114 mm)

General description

Tefzel® Fluorocarbon plastic forceps have smooth square tips 1.6mm (1/16") wide.

Features and Benefits

  • Excellent for handling small balance weights
  • Will withstand temperature up to 199ºC (390ºF); autoclavable
  • Individually blister packed

Legal Information

Bel-Art is a registered trademark of SP Scienceware, a division of SP
Tefzel is a registered trademark of E. I. du Pont de Nemours and Company

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service