Key Documents
BAH182140000
SP Bel-Art® Plastic Spring Style Tubing Clamps
for tubing o.d., up to 5/32 in. (12 mm)
Select a Size
3 EACH
$116.00
$116.00
Check Cart for Availability
Select a Size
Change View
3 EACH
$116.00
About This Item
UNSPSC Code:
41122800
NACRES:
NB.43
$116.00
Check Cart for Availability
feature
for tubing o.d. up to 5/32 in. (12 mm)
packaging
pack of 3 ea
manufacturer/tradename
Bel-Art H18214-0000
General description
Perfect for all flexible tubing, this positive-action, self-closing tube clamp opens by squeezing the handles together.
Features and Benefits
  • Chemical and stain-resistant
  • Made of Acetal plastic and stainless steel
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)