Merck
All Photos(4)
Key Documents
BAF371250012
Spinfin® magnetic stirring bar
H × diam. 0.5 in. (12.7 mm) × 0.5 in. (12.7 mm)
Synonym(s):
Spinfin® magnetic stirring bar
About This Item
UNSPSC Code:
41122800
NACRES:
NB.51
Pricing and availability is not currently available.
material
PTFE-coated
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F37125-0012
H × diam.
0.5 in. (12.7 mm) × 0.5 in. (12.7 mm)
General description
Disks have fins on top for complete agitation.
Spinfin® magnetic stirring bars agitate liquids in round bottom vessels such as test tubes, cylinders and flasks, where other stirring bars cannot be used.
Features and Benefits
  • Maintains its position in a vessel and will not wobble
  • Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE coating – Superior Chemical Compatibility
  • Manufactured in a registered ISO 9001:2015 facility, as verified by SGS certification
  • Autoclave safe
Legal Information
Spinfin is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)