BAF371220035
SP Bel-Art® Spinbar® magnetic stir bar
polygon white PTFE-coated, L × diam. 1.375 in. (35 mm) × 0.25 in. (6 mm)
Synonym(s):
Spinbar® magnetic stir bar
Select a Size
$17.20
Estimated to ship on16 April 2025
Select a Size
About This Item
$17.20
Estimated to ship on16 April 2025
Recommended Products
material
polygon white PTFE-coated
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F37122-0035
L × diam.
1.375 in. (35 mm) × 0.25 in. (6 mm)
General description
- Spinbar® magnetic stir bar is used in conjunction with a magnetic stirring plate for uniform, consistent, and thorough mixing of chemical solutions.
- Alnico magnet encapsulated in durable and inert FDA grade polytetrafluoroethylene (PTFE) low friction coating.
- Multi-faceted-shaped bars provide a greater surface area and added turbulence for more vigorous, thorough mixing.
- Autoclave safe.
Features and Benefits
- Center molded pivot ring minimizes the contact area between the bar and the vessel; reduces friction and chattering
- Marring of plastic containers is diminished
- Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE coating – Superior Chemical Compatibility
- Manufactured in a registered ISO 9001:2015 facility, as verified by SGS certification
- Autoclave safe
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)