BAF371110138
SP Bel-Art® Spinbar® magnetic stir bar
cylindrical white PTFE-coated, L × diam. 1 in. (25.4 mm) × 0.375 in. (9.5 mm)
Synonym(s):
Spinbar® magnetic stir bar
Select a Size
$17.20
Estimated to ship on10 April 2025
Select a Size
About This Item
$17.20
Estimated to ship on10 April 2025
Recommended Products
material
cylindrical white PTFE-coated
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F37111-0138
L × diam.
1 in. (25.4 mm) × 0.375 in. (9.5 mm)
Related Categories
General description
- Spinbar® magnetic stir bar is used in conjunction with a magnetic stirring plate for uniform, consistent, and thorough mixing of chemical solutions.
- Alnico magnet encapsulated in durable and inert FDA grade polytetrafluoroethylene (PTFE) low friction coating.
- Multi-faceted-shaped bars provide a greater surface area and added turbulence for more vigorous, thorough mixing.
- Autoclave safe.
Features and Benefits
- Small removable pivot ring in the center adds to their versatility
- Pivot ring minimizes the contact area of the bar to the vessel, reduces friction and lessens marring of plastic containers
- Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE coating – Superior Chemical Compatibility
- Manufactured in a registered ISO 9001:2015 facility, as verified by SGS certification
- Autoclave safe
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)