Merck
All Photos(4)
Key Documents
BAF369751000
SP Bel-Art® Sterileware Powder Probe Samplers
white high-density polyethylene, capacity 200 mL (6.8 oz), L 38 in. (96.5 cm), sterile, pack of 20 ea (individually wrapped)
Synonym(s):
sampler
About This Item
UNSPSC Code:
41122800
NACRES:
NB.35
Pricing and availability is not currently available.
material
white high-density polyethylene
sterility
sterile
packaging
pack of 20 ea (individually wrapped)
manufacturer/tradename
Bel-Art F36975-1000
L
38 in. (96.5 cm)
capacity
200 mL (6.8 oz)
General description
Ideal for taking a cross sectional sample through a bulk powder, this probe is available in two convenient lengths. Used by pharmaceutical, food, cosmetic and chemical companies.
Features and Benefits
  • End tip contains free-flowing powders within the probe
  • Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
  • High density polyethylene (HDPE) probe is heat sealed in individual polyethylene bag
  • Gamma irradiated sterile (SAL 10^-6)Ready for one time use and disposal
  • Lot stamped for accurate tracing Note: Sterilized products are not returnable
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Sterileware is a trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)