Skip to Content
Merck
All Photos(1)

Key Documents

BAF116460628

Scienceware® safety labeled wash bottles

label, Toluene, wide-mouth red cap, capacity 500 mL (16 oz)

Synonym(s):

dispensing bottle, safety bottles, squeeze bottle

Sign Into View Organizational & Contract Pricing

Select a Size

6 EACH
$87.90

$87.90


Check Cart for Availability


Select a Size

Change View
6 EACH
$87.90

About This Item

UNSPSC Code:
41122800
NACRES:
NB.43

$87.90


Check Cart for Availability

material

low-density polyethylene
wide-mouth red cap

feature

label Toluene

packaging

pack of 6 ea

manufacturer/tradename

Bel-Art F11646-0628

capacity

500 mL
500 mL (16 oz)

color

opaque

Looking for similar products? Visit Product Comparison Guide

General description

Bel-Art’s Safety Labeled wash bottles include Department of Transportation (DOT) and National Fire Protection Association (NFPA) codes and symbols, Chemical Abstract Service (CAS) number, protective equipment symbols and target organ information, making your workplace a safer place.
Safety Labeled wash bottles with Department of Transportation (DOT) and National Fire Protection Association (NFPA) codes and symbols, Chemical Abstract Service (CAS) number, protective equipment symbols and target organ information, for making your workplace a safer place.

Features and Benefits

  • Easy-to-read printed bottles
  • Wide mouth 53mm (2") bottle for easy refilling
  • Translucent, LDPE solvent-resistant; Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm; Toluene is red LDPE bottle
  • Polypropylene closures
  • Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
Note: Use LYOB only with solvents compatible with LDPE.
  • Easy-to-read printed bottles
  • Wide mouth 53mm (2") bottle for easy refilling
  • Translucent, LDPE solvent-resistant; Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm; Toluene is red LDPE bottle
  • Polypropylene closures
  • Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200

Legal Information

Bel-Art is a registered trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service