Skip to Content
Merck
All Photos(1)

Key Documents

B89507

Sigma-Aldrich

2-(Butylamino)ethanethiol

97%

Synonym(s):

2-(Butylamino)ethane-1-thiol, 2-n-Butylaminoethanethiol

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$412.00

$412.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$412.00

About This Item

Linear Formula:
CH3(CH2)3NHCH2CH2SH
CAS Number:
Molecular Weight:
133.26
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$412.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.4711 (lit.)

bp

112-115 °C/10 mmHg (lit.)

density

0.901 g/mL at 25 °C (lit.)

SMILES string

CCCCNCCS

InChI

1S/C6H15NS/c1-2-3-4-7-5-6-8/h7-8H,2-6H2,1H3

InChI key

OUMFAUYLXGTBCX-UHFFFAOYSA-N

Application

  • Photocatalytic oxidation of VX simulant 2-(butylamino)ethanethiol.: This paper investigates the photocatalytic oxidation of 2-(Butylamino)ethanethiol, a simulant for the nerve agent VX. The findings indicate that photocatalytic processes can effectively degrade sulfur-containing compounds, suggesting applications in decontamination and hazardous material management. This research provides insights into the environmental remediation of toxic chemical agents (Vorontsov et al., 2004).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

163.4 °F - closed cup

Flash Point(C)

73 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service