Skip to Content
Merck
All Photos(3)

Key Documents

B84505

Sigma-Aldrich

Butadiene sulfone

98%

Synonym(s):

2,5-Dihydrothiophene-1,1-dioxide, 3-Sulfolene

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$59.00
500 G
$108.00

$59.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$59.00
500 G
$108.00

About This Item

Empirical Formula (Hill Notation):
C4H6O2S
CAS Number:
Molecular Weight:
118.15
Beilstein:
107004
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$59.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

crystals

mp

65-66 °C (lit.)

SMILES string

O=S1(=O)CC=CC1

InChI

1S/C4H6O2S/c5-7(6)3-1-2-4-7/h1-2H,3-4H2

InChI key

MBDNRNMVTZADMQ-UHFFFAOYSA-N

General description

Butadiene sulfone is a β, γ-unsaturated cyclic sulfone that can switch from solvent to 1,3-butadiene and sulfur dioxide in the presence of water. It is commonly used as a building block especially in the synthesis of functionalized dienes, through alkylation, deprotonation, and cheletropic SO2 removal. [1] [2]

Application

  • Stretchable PEDOT:PSS/Li-TFSI/XSB Composite Films: Butadiene sulfone plays a critical role in the synthesis of composite films designed for electromagnetic interference shielding, indicating its potential in the production of advanced electronic materials and contributing to the development of stretchable electronics (Jiang et al., 2023).
  • Applications of 3D printed bone tissue engineering scaffolds: Research underscores the versatility of butadiene sulfone as a solvent in the 3D printing of bone tissue scaffolds, emphasizing its utility in regenerative medicine and the manufacturing of biocompatible engineering materials (Su et al., 2021).
  • Zwitterionic phosphonium ligands: The study highlights the synthesis and application of zwitterionic phosphonium ligands using butadiene sulfone, showcasing its efficacy in the telomerization process of 1,3-butadiene, which is essential for the production of specialty chemicals (Pews-Davtyan et al., 2016).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

233.6 °F - closed cup

Flash Point(C)

112 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Switchable butadiene sulfone pretreatment of Miscanthus in the presence of water
J Atilio De, et al.
Green Chemistry, 15, 1067-1078 (2013)
Tandem vinylogous 1, 2-addition/anionic oxy-Cope reaction leading from butadiene sulfone to an orthogonally functionalized bicycle
Michael B G, et al.
The Journal of Organic Chemistry, 75, 6312-6315 (2010)
Monika Johansson et al.
Journal of pharmaceutical and biomedical analysis, 100, 215-229 (2014-08-30)
An effective screening procedure to identify and quantify active pharmaceutical substances in suspected illegal medicinal products is described. The analytical platform, consisting of accurate mass determination with liquid chromatography time-of-flight mass spectrometry (LC-QTOF-MS) in combination with nuclear magnetic resonance (NMR)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service