Skip to Content
Merck
All Photos(1)

Key Documents

B83800

Sigma-Aldrich

4-Bromo-1,2-dimethylbenzene

70%

Synonym(s):

4-Bromo-o-xylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H3Br
CAS Number:
Molecular Weight:
185.06
Beilstein:
2040354
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

70%

form

liquid

refractive index

n20/D 1.555 (lit.)

bp

215 °C (lit.)

density

1.37 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(Br)cc1C

InChI

1S/C8H9Br/c1-6-3-4-8(9)5-7(6)2/h3-5H,1-2H3

InChI key

QOGHRLGTXVMRLM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

remainder 3-bromo-o-xylene

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

176.0 °F - closed cup

Flash Point(C)

80 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Arivazhagan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 91, 419-430 (2012-03-21)
In the present work, we reported a combined experimental and theoretical study on molecular structure, vibrational spectra and NBO analysis of 4-bromo-o-xylene (BOX). The FT-IR (400-4000 cm(-1)) and FT-Raman spectra (50-3500 cm(-1)) of BOX were recorded. The molecular geometry, harmonic
[Mechanism of disorders of reproductive function after exposure to 4-brom-o-xylol (experimental studies)].
M A Fesenko
Gigiena truda i professional'nye zabolevaniia, (3)(3), 48-49 (1989-01-01)
V V Iaglov et al.
Gigiena truda i professional'nye zabolevaniia, (5)(5), 23-25 (1991-01-01)
The article contains a comparative analysis of experimental data on the assessment of the general toxicological, gonadotrophic effects and morphofunctional reactions of tissue basophils under the influence of 4-bromine-o-xylene, carbon disulfide and pyracryl. Established was the gonadotrophic action of pyracryl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service