Skip to Content
Merck
All Photos(1)

Documents

B79684

Sigma-Aldrich

2-Bromopropiophenone

technical grade, 90%

Synonym(s):

α-Bromopropiophenone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COCHBrCH3
CAS Number:
Molecular Weight:
213.07
Beilstein:
508550
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

90%

form

liquid

refractive index

n20/D 1.571 (lit.)

bp

245-250 °C (lit.)

density

1.4 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(Br)C(=O)c1ccccc1

InChI

1S/C9H9BrO/c1-7(10)9(11)8-5-3-2-4-6-8/h2-7H,1H3

InChI key

WPDWOCRJBPXJFM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service