Skip to Content
Merck
All Photos(1)

Key Documents

B74305

Sigma-Aldrich

2-Bromo-2-nitropropane

97%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$200.00

$200.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$200.00

About This Item

Linear Formula:
BrC(CH3)2NO2
CAS Number:
Molecular Weight:
167.99
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$200.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.464 (lit.)

bp

48 °C/15 mmHg (lit.)

density

1.582 g/mL at 25 °C (lit.)

SMILES string

CC(C)(Br)[N+]([O-])=O

InChI

1S/C3H6BrNO2/c1-3(2,4)5(6)7/h1-2H3

InChI key

OADSZWXMXIWZSQ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

141.8 °F - closed cup

Flash Point(C)

61 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N L Whitehouse et al.
Journal of dairy science, 100(12), 9585-9601 (2017-10-02)
Estimates of Lys bioavailability of rumen-protected Lys (RP-Lys) supplements are often obtained using in vitro or 2-step in situ techniques, with little to no data determining efficacy and bioavailability in vivo. The objective of this study was to further evaluate
D L Morris et al.
Journal of dairy science, 103(8), 7110-7123 (2020-06-09)
Hydrolyzed feather meal (HFM) is high in crude protein, most of which bypasses rumen degradation when fed to lactating dairy cows, allowing direct supply of AA to the small intestine. Compared with other feeds that are high in bypass protein

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service