Skip to Content
Merck
All Photos(1)

Documents

B28656

Sigma-Aldrich

5-Benzyloxytryptamine hydrochloride

98%

Synonym(s):

5-(phenylmethoxy)indole-3-ethanamine monohydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H18N2O · HCl
CAS Number:
Molecular Weight:
302.80
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

mp

260-263 °C (lit.)

SMILES string

Cl[H].NCCc1c[nH]c2ccc(OCc3ccccc3)cc12

InChI

1S/C17H18N2O.ClH/c18-9-8-14-11-19-17-7-6-15(10-16(14)17)20-12-13-4-2-1-3-5-13;/h1-7,10-11,19H,8-9,12,18H2;1H

InChI key

IUWVJCIEWSQGHH-UHFFFAOYSA-N

Application

Reactant for preparation of:
  • Indole alkaloid-based compounds for drug discovery
  • Serotonin 5-HT6 receptor ligands

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Maki Tsumura et al.
PloS one, 8(12), e82233-e82233 (2013-12-21)
Odontoblasts produce dentin during development, throughout life, and in response to pathological conditions by sensing stimulation of exposed dentin. The functional properties and localization patterns of transient receptor potential (TRP) melastatin subfamily member 8 (TRPM8) and ankyrin subfamily member 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service