Skip to Content
Merck
All Photos(2)

Key Documents

B27803

Sigma-Aldrich

5-(Benzyloxy)indole

95%

Synonym(s):

NSC 62895

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$50.10
25 G
$134.00

$50.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$50.10
25 G
$134.00

About This Item

Empirical Formula (Hill Notation):
C15H13NO
CAS Number:
Molecular Weight:
223.27
Beilstein:
173532
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$50.10


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

powder

impurities

5% various solvents of crystallization

mp

100-104 °C (lit.)

SMILES string

C(Oc1ccc2[nH]ccc2c1)c3ccccc3

InChI

1S/C15H13NO/c1-2-4-12(5-3-1)11-17-14-6-7-15-13(10-14)8-9-16-15/h1-10,16H,11H2

InChI key

JCQLPDZCNSVBMS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Reactant in regio- and stereoselective morpholine-catalyzed direct C-3 alkenylation with α,β-unsaturated aldehydes
  • Reactant in selective debenzylation of protective groups using SiliaCat-palladium under mild reaction conditions
  • Reactant in metal-free Friedel-Crafts alkylation reactions
  • Reactant in preparation of protein kinase (PKC) inhibitors
  • Reactant in preparation of indole/quinoline carbothioic acid amide derivatives

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jeff DeFalco et al.
Bioorganic & medicinal chemistry letters, 20(23), 7076-7079 (2010-10-23)
5-Benzyloxytryptamine 19 was found to act as an antagonist of the TRPM8 ion-channel. For example, 19 had an IC(50) of 0.34 μM when menthol was used as the stimulating agonist. Related commercially-available tryptamine derivatives showed diminished, or no antagonist activity

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service