All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C12H9NO
CAS Number:
Molecular Weight:
183.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Recommended Products
Assay
97%
bp
307 °C (lit.)
mp
36-40 °C (lit.)
SMILES string
O=C(c1ccccc1)c2cccnc2
InChI
1S/C12H9NO/c14-12(10-5-2-1-3-6-10)11-7-4-8-13-9-11/h1-9H
InChI key
RYMBAPVTUHZCNF-UHFFFAOYSA-N
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
302.0 °F - closed cup
Flash Point(C)
150 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
P Eyer et al.
Xenobiotica; the fate of foreign compounds in biological systems, 13(11), 649-659 (1983-11-01)
3-Benzoylpyridine (3-BP), a decomposition product of the soman antidote, HGG-12 (3-benzoylpyridino(1)-methyl 2'-hydroxyiminomethylpyridino(1')methyl ether dichloride) was rapidly metabolized in the isolated perfused rat liver, giving 3-(alpha-hydroxybenzyl)pyridine and its corresponding glucuronide, 3-benzoylpyridine-N-oxide, and 3-(alpha-hydroxybenzyl)pyridine-N-oxide. The latter is formed both from 3-(alpha-hydroxybenzyl)pyridine and
M Murray et al.
Chemico-biological interactions, 113(3), 161-173 (1998-08-26)
3-Benzoylpyridine (3BP) is a major metabolite of HGG-12, and oxime that has been synthesized as a potential antidote to the toxic effects of soman and other anticholinesterases. Structural similarities exist between 3BP, the cytochrome P450 (CYP)-inducer metyrapone (MET) and other
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service