Skip to Content
Merck
All Photos(1)

Key Documents

B12601

Sigma-Aldrich

4-Benzoylbiphenyl

99%

Synonym(s):

4-Phenylbenzophenone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H4COC6H5
CAS Number:
Molecular Weight:
258.31
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

powder

bp

419-420 °C (lit.)

mp

99-101 °C (lit.)

SMILES string

O=C(c1ccccc1)c2ccc(cc2)-c3ccccc3

InChI

1S/C19H14O/c20-19(17-9-5-2-6-10-17)18-13-11-16(12-14-18)15-7-3-1-4-8-15/h1-14H

InChI key

LYXOWKPVTCPORE-UHFFFAOYSA-N

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryohei Shimizu et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 17(9), 1206-1212 (2018-08-01)
Diethylhexyl 2,6-naphthalate (DEHN, tradename Corapan TQ) is used as a photostabilizer for a widely-used UV-A absorber, butylmethoxydibenzoylmethane (BMDBM). The photophysical properties of DEHN have been studied through measurements of the transient absorption, fluorescence, phosphorescence and EPR spectra in ethanol. The
K Van Den Houwe et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 34(7), 1261-1269 (2017-05-18)
Contaminants in food packaging are a challenge of our time since the packaging material itself has been found to represent a source of food contamination through the migration of substances from it. Before first use, packaging materials destined for the
Juan Li et al.
Environmental science & technology, 53(18), 10969-10977 (2019-08-15)
Photoinitiators (PIs) are widely used in industrial polymerization and have been detected as emerging contaminants in environmental matrixes. It has been reported that humans are exposed to PIs, but the maternal-fetal transmission of PIs has not been documented. In this
Miguel A Lago et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 33(3), 518-529 (2016-01-21)
Since the UV ink photoinitiator (PI) isopropylthioxanthone (ITX) was discovered in packaged milk, studies of print contamination have focused primarily on PIs but have also included amine synergists. Many other substances are used or formed during the print process, yet
Coraline Simon et al.
Toxicology in vitro : an international journal published in association with BIBRA, 37, 121-133 (2016-10-19)
Endocrine activity of 65 compounds migrating from polycarbonate replacement plastic baby bottles was assessed using in vitro cell based assays (reporter gene assays) involving 7 nuclear receptors, i.e. human steroid hormones receptors (oestrogen, androgen, progesterone and glucocorticoid receptors), human thyroid

Protocols

Thrombin is an endolytic serine protease that selectively cleaves the Arg–Gly bonds of fibrinogen to form fibrin and release fibrinopeptides A and B.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service