Skip to Content
Merck
All Photos(2)

Key Documents

ALD00615

Sigma-Aldrich

Ethyl (S,E)-2-((mesitylsulfinyl)imino)acetate

≥95%

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$664.00

$664.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$664.00

About This Item

Empirical Formula (Hill Notation):
C13H17NO3S
CAS Number:
Molecular Weight:
267.34
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

$664.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95%

form

powder

reaction suitability

reaction type: C-C Bond Formation

storage temp.

−20°C

SMILES string

[S](=O)(\N=C\C(=O)OCC)c1c(cc(cc1C)C)C

InChI

1S/C13H17NO3S/c1-5-17-12(15)8-14-18(16)13-10(3)6-9(2)7-11(13)4/h6-8H,5H2,1-4H3/b14-8+

InChI key

OUMNTVUUJPMNPJ-RIYZIHGNSA-N

Application

Ethyl (S,E)-2-((mesitylsulfinyl)imino)acetate is a chiral glyoxylate-derived sulfinimine reagent used for the synthesis of optically pure amino acids through a decarboxylative radical cross-coupling reaction.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shengyang Ni et al.
Angewandte Chemie (International ed. in English), 57(44), 14560-14565 (2018-09-14)
The direct union of primary, secondary, and tertiary carboxylic acids with a chiral glyoxylate-derived sulfinimine provides rapid access into a variety of enantiomerically pure α-amino acids (>85 examples). Characterized by operational simplicity, this radical-based reaction enables the modular assembly of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service