Skip to Content
Merck
All Photos(1)

Key Documents

ALD00442

Sigma-Aldrich

Sodium butane-1-sulfinate

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$377.00

$377.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$377.00

About This Item

Empirical Formula (Hill Notation):
C4H9NaO2S
Molecular Weight:
144.17
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

$377.00


Check Cart for Availability

Request a Bulk Order

form

powder

Quality Level

reaction suitability

reaction type: C-C Bond Formation

mp

250 °C

SMILES string

CCCCS(O[Na])=O

Application

The following Baran Sulfinate is a part of a toolbox of diversification reagents, which are ideal for the late-stage functionalization of nitrogen-containing heterocycles. The regioselectivity can be effectively tuned by modification of the pH and solvent selection.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service