Skip to Content
Merck
All Photos(2)

Key Documents

ALD00352

Sigma-Aldrich

Boc-2-methoxy-L-phenylalanine

97%

Synonym(s):

(S)-2-(tert-butoxycarbonylamino)-3-(2-methoxyphenyl)propanoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$440.00

$440.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$440.00

About This Item

Empirical Formula (Hill Notation):
C15H21NO5
CAS Number:
Molecular Weight:
295.33
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

$440.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

powder

reaction suitability

reaction type: Boc solid-phase peptide synthesis
reagent type: ligand
reaction type: C-H Activation

mp

157 °C

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

OC([C@@H](NC(OC(C)(C)C)=O)CC1=CC=CC=C1OC)=O

InChI

1S/C15H21NO5/c1-15(2,3)21-14(19)16-11(13(17)18)9-10-7-5-6-8-12(10)20-4/h5-8,11H,9H2,1-4H3,(H,16,19)(H,17,18)/t11-/m0/s1

InChI key

QMHKMTAKTUUKEK-NSHDSACASA-N

Application

Unnatural amino acid was derived from a C-H Activation methodology developed by Jin-Quan Yu and coworkers. The Pd-mediated C-C bond formation can be made in tandem with 2-Methylpyridine (Aldrich 109835).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service