Skip to Content
Merck
All Photos(1)

Key Documents

ALD00198

Sigma-Aldrich

3-Chloro-4-methylbenzenesulfonyl fluoride

95% (GC)

Synonym(s):

3-Chloro-4-methylbenzenesulfonyl fluoride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6ClFO2S
CAS Number:
Molecular Weight:
208.64
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95% (GC)

form

liquid

reaction suitability

reaction type: click chemistry

refractive index

n/D 1.5165

density

1.3992 at 25 °C

SMILES string

ClC1=C(C)C=CC(S(=O)(F)=O)=C1

InChI

1S/C7H6ClFO2S/c1-5-2-3-6(4-7(5)8)12(9,10)11/h2-4H,1H3

InChI key

GMIYUPVDARXJAI-UHFFFAOYSA-N

Application

Sulfonyl fluoride motif can be used as a connector for the assembly of -SO2- linked small molecules with proteins or nucleic acids. This new click chemistry approach through sulfates is a complimentary approach to using amides and phosphate groups as linkers.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service