Skip to Content
Merck
All Photos(1)

Key Documents

A44150

Sigma-Aldrich

4-Aminobutyraldehyde diethyl acetal

90%, technical grade

Synonym(s):

4,4-Diethoxybutylamine

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$120.00
25 G
$299.00
100 G
$976.00

$120.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$120.00
25 G
$299.00
100 G
$976.00

About This Item

Linear Formula:
H2N(CH2)3CH(OC2H5)2
CAS Number:
Molecular Weight:
161.24
Beilstein:
1739649
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$120.00


Check Cart for Availability

Request a Bulk Order

grade

technical grade

Quality Level

Assay

90%

form

liquid

reaction suitability

reagent type: cross-linking reagent

refractive index

n20/D 1.428 (lit.)

bp

196 °C (lit.)

density

0.933 g/mL at 25 °C (lit.)

SMILES string

CCOC(CCCN)OCC

InChI

1S/C8H19NO2/c1-3-10-8(11-4-2)6-5-7-9/h8H,3-7,9H2,1-2H3

InChI key

GFLPSABXBDCMCN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eva Ternon et al.
Molecules (Basel, Switzerland), 24(1) (2019-01-04)
Due to increasing evidence of key chemically mediated interactions in marine ecosystems, a real interest in the characterization of the metabolites involved in such intra and interspecific interactions has emerged over the past decade. Nevertheless, only a small number of
Dana Al Sulaiman et al.
Nano letters, 18(9), 6084-6093 (2018-08-15)
Label-free, single-molecule sensing is anideal candidate for biomedical applications that rely on the detection of low copy numbers in small volumes and potentially complex biofluids. Among them, solid-state nanopores can be engineered to detect single molecules of charged analytes when
Janchai Poonlaphdecha et al.
Food chemistry, 197(Pt A), 965-971 (2015-12-01)
The role of 1-pyrroline was studied via feeding experiments using rice calli cultures to gain further insight into the key steps of 2-acetyl-1-pyrroline (2AP) biosynthesis in rice. The origin of the acetyl donor was also studied through stable isotope labelled
Yang Yang et al.
Journal of the American Society of Nephrology : JASN, 29(9), 2310-2325 (2018-07-26)
Autosomal-dominant polycystic kidney disease (ADPKD) is the leading inherited renal disease worldwide. The proproliferative function of macrophages is associated with late-stage cyst enlargement in mice with PKD; however, the way in which macrophages act on cyst-lining epithelial cells (CLECs) has
Carlos Eduardo Flores-Tinoco et al.
Molecular systems biology, 16(6), e9419-e9419 (2020-06-04)
Biological nitrogen fixation emerging from the symbiosis between bacteria and crop plants holds promise to increase the sustainability of agriculture. One of the biggest hurdles for the engineering of nitrogen-fixing organisms is an incomplete knowledge of metabolic interactions between microbe

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service