Skip to Content
Merck
All Photos(1)

Key Documents

96823

Sigma-Aldrich

tert-Butyl trans-17-bromo-4,7,10,13-tetraoxa-15-heptadecenoate

≥95.0% (AT)

Synonym(s):

Hycron linker

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H31BrO6
CAS Number:
Molecular Weight:
411.33
Beilstein:
7214333
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥95.0% (AT)

form

liquid

reaction suitability

reagent type: cross-linking reagent

color

brown

refractive index

n20/D 1.475

density

1.182 g/mL at 20 °C (lit.)

SMILES string

CC(C)(C)OC(=O)CCOCCOCCOCCOC\C=C\CBr

InChI

1S/C17H31BrO6/c1-17(2,3)24-16(19)6-9-21-11-13-23-15-14-22-12-10-20-8-5-4-7-18/h4-5H,6-15H2,1-3H3/b5-4+

InChI key

ZCDCGWNGIROULA-SNAWJCMRSA-N

Other Notes

HYCRON, an allylic anchor for high-efficiency solid phase synthesis of protected peptides and glycopeptides

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O. Seitz, H. Kunz
The Journal of Organic Chemistry, 62, 813-813 (1997)
O. Seitz et al.
Angewandte Chemie (International Edition in English), 111, 2340-2340 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service