Skip to Content
Merck
All Photos(1)

Key Documents

96083

Sigma-Aldrich

Z-Dab-OH

≥99.0% (sum of enantiomers, TLC)

Synonym(s):

Nα-Z-L-2,4-diaminobutyric acid, Z-Dbu-OH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H16N2O4
CAS Number:
Molecular Weight:
252.27
Beilstein:
2288043
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥99.0% (sum of enantiomers, TLC)

optical activity

[α]20/D −9.5±0.5°, c = 1% in H2O

mp

194-196 °C

application(s)

peptide synthesis

SMILES string

NCC[C@H](NC(=O)OCc1ccccc1)C(O)=O

InChI

1S/C12H16N2O4/c13-7-6-10(11(15)16)14-12(17)18-8-9-4-2-1-3-5-9/h1-5,10H,6-8,13H2,(H,14,17)(H,15,16)/t10-/m0/s1

InChI key

KXMSCBRTBLPGIN-JTQLQIEISA-N

Other Notes

Mono-protected derivative of L-2,4-diaminobutyric acid; used e.g. for the synthesis of α-amino-γ-guanidino butyric acid; synthesis of an analog of virginiamycin

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M.J.O. Anteunis et al.
Bull. Soc. Chim. Belg., 97, 135-135 (1988)
F. Brtnik et al.
Collection of Czechoslovak Chemical Communications, 41, 2969-2969 (1976)
H. Arold et al.
J. Prakt. Chem., 317, 682-682 (1975)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service