Skip to Content
Merck
All Photos(1)

Key Documents

95517

Sigma-Aldrich

4-Bromo-1-butanol

technical, ≥85% (as solvent-free, GC)

Synonym(s):

Tetramethylene bromohydrin

Sign Into View Organizational & Contract Pricing

Select a Size

2.5 G
$220.00

$220.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
2.5 G
$220.00

About This Item

Linear Formula:
BrCH2CH2CH2CH2OH
CAS Number:
Molecular Weight:
153.02
Beilstein:
1839430
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$220.00


Check Cart for Availability

Request a Bulk Order

grade

technical

Assay

≥85% (as solvent-free, GC)

form

liquid

functional group

bromo
hydroxyl

storage temp.

−20°C

SMILES string

OCCCCBr

InChI

1S/C4H9BrO/c5-3-1-2-4-6/h6H,1-4H2

InChI key

SIJLYRDVTMMSIP-UHFFFAOYSA-N

Analysis Note

contains varying amounts of tetrahydrofuran as solvent

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

44.6 °F

Flash Point(C)

7 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiang Xie et al.
Scientific reports, 7(1), 15735-15735 (2017-11-18)
The influence of the linking group between the phosphoryl and bridging moieties in phosphoryl-containing task-specific ionic liquids (TSILs) on the extraction of uranyl ions was experimentally and theoretically investigated. A novel phosphoramidate-based TSIL with an amine group as the linking

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service