Skip to Content
Merck
All Photos(1)

Documents

936103

Sigma-Aldrich

tBuXPhos Pd G6 Br

Synonym(s):

tBuXphos OAC precatalyst Br

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H55BrNO2PPd
Molecular Weight:
823.19
UNSPSC Code:
12161600
NACRES:
NA.21

form

powder

Quality Level

SMILES string

Br[Pd-](C1=CC=C(NC(OC2=CC=CC=C2)=O)C=C1)[P+](C(C=CC=C3)=C3C(C(C(C)C)=CC(C(C)C)=C4)=C4C(C)C)(C(C)(C)C)C(C)(C)C

Application

tBuXPhos Pd G6 Br is an oxadative addition complex (OAC) of tBuXPhos for use in bioconjugation. The OAC can be used to arylate cysteine followed by subsequent protein-peptide cross-linking.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service