Skip to Content
Merck
All Photos(2)

Key Documents

935654

Sigma-Aldrich

Tris(trimethylsilyl) borate

new
greener alternative

≥99.5%, battery grade

Synonym(s):

Boric acid tris(trimethylsilyl) ester, TMSB, Tris(trimethylsiloxy)borane, Tris(trimethylsiloxy)boron

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3SiO]3B
CAS Number:
Molecular Weight:
278.38
Beilstein:
1772733
MDL number:
UNSPSC Code:
12352005
Assay:
≥99.5%
grade:
battery grade
form:
(Colorless liquid)

grade

battery grade

Quality Level

description

Application: Battery manufacturing

Assay

≥99.5%

form

(Colorless liquid)

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.386 (lit.)

bp

186 °C (lit.)
186 °C

mp

-35 °C

density

0.831 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

greener alternative category

SMILES string

C[Si](C)(C)OB(O[Si](C)(C)C)O[Si](C)(C)C

InChI

1S/C9H27BO3Si3/c1-14(2,3)11-10(12-15(4,5)6)13-16(7,8)9/h1-9H3

InChI key

YZYKZHPNRDIPFA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tris(trimethylsilyl) borate (TMSB) is an electron-deficient organoborate compound. It is a colorless liquid that that is miscible with many organic solvents, including the linear and cyclic carbonates used in battery applications. Our battery grade TMSB is differentiated by its high purity and low trace water content to ensure consistent and reliable performance in batteries.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more information.

Application

Tris(trimethylsilyl) borate has a wide range of applications in various industries, including battery manufacturing, where it is used as an electrolyte additive, particularly in high-voltage lithium-ion batteries. TMSB is a cathode-film-forming additive that forms a thin protective coating called a solid-electrolyte interface (SEI) on the surface of the cathode. Adding TMSB to the electrolyte formulation, even at low concentrations of 0.5 wt.%, noticeably improves capacity retention for NMC, LMO, and LMNO cathodes. TMSB also is shown to reduce the flammability of the electrolyte and improve the thermal stability of LiPF6-based electrolytes at elevated temperatures.
TMSB also has catalytic properties, which make it useful in organic synthesis reactions and as a reagent in the manufacturing of pharmaceuticals and other chemicals.

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

105.8 °F - closed cup

Flash Point(C)

41 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tris(trimethylsilyl)borate as an electrolyte additive for improving interfacial stability of high voltage layered lithium-rich oxide cathode/carbonate-based electrolyte
Li J, et al
Journal of Power Sources, 285, 360-366 (2015)
Effect of tris(trimethylsilyl)borate on the high voltage capacity retention of LiNi0.5Co0.2Mn0.3O2/graphite cells
Zuo X, et al.
Journal of Power Sources, 229, 308-312 (2013)
Tris(trimethylsilyl) borate as an electrolyte additive to improve the cyclability of LiMn2O4cathode for lithium-ion battery
Liu Y, et al.
Journal of Power Sources, 221, 90-96 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service