Skip to Content
Merck
All Photos(1)

Key Documents

934321

Sigma-Aldrich

L-Valyl-L-citrulline

≥95%

Synonym(s):

L-Valyl-N5-(aminocarbonyl)-L-ornithine, Val-Cit

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$374.00

$374.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$374.00

About This Item

Empirical Formula (Hill Notation):
C11H22N4O4
CAS Number:
Molecular Weight:
274.32
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.21

$374.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95%

form

powder or crystals

color

off-white to light yellow

storage temp.

2-8°C

Application

This is a cathepsin cleavable linker used in the construction of antibody drug conjugates (ADC & Bioconjugation).

Features and Benefits

Val-Cit-OH is a cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADC). The Val-Cit is specifically cleaved by cathepsin B. The Boc group can be deprotected under acidic conditions to generate a free amine group. As this enzyme is only present in the lysosome the ADC payload will be release only in the cell.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qinhuai Lai et al.
European journal of medicinal chemistry, 199, 112364-112364 (2020-05-14)
Cryptophycin-52 (CR52), a tubulin inhibitor, exhibits promising antitumor activity in vitro (picomolar level) and in mouse xenograft models. However, the narrow therapeutic window in clinical trials limits its further development. Antibody-drug conjugate (ADC), formed by coupling cytotoxic compound (payload) to an
Zixuan Ye et al.
International journal of nanomedicine, 16, 2443-2459 (2021-04-06)
Specific modifications to carriers to achieve targeted delivery of chemotherapeutics into malignant tissues are a critical point for efficient diagnosis and therapy. In this case, bovine serum albumin (BSA) was conjugated with cetuximab-valine-citrulline (vc)-doxorubicin (DOX) to target epidermal growth factor

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service