Skip to Content
Merck
All Photos(1)

Key Documents

93404

Sigma-Aldrich

Tris(N,N-tetramethylene)phosphoric acid triamide

≥98.0% (GC)

Synonym(s):

Phosphoric acid tripyrrolidide, Tripyrrolidinophosphine oxide

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$254.00
25 ML
$1,030.00

$254.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
$254.00
25 ML
$1,030.00

About This Item

Empirical Formula (Hill Notation):
C12H24N3OP
CAS Number:
Molecular Weight:
257.31
Beilstein:
3550018
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$254.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98.0% (GC)

form

liquid

refractive index

n20/D 1.514

bp

140-142 °C/0.1 mmHg (lit.)

density

1.120 g/mL at 20 °C (lit.)

SMILES string

O=P(N1CCCC1)(N2CCCC2)N3CCCC3

InChI

1S/C12H24N3OP/c16-17(13-7-1-2-8-13,14-9-3-4-10-14)15-11-5-6-12-15/h1-12H2

InChI key

GQOGEHIVQIMJMO-UHFFFAOYSA-N

Caution

may darken on storage

Other Notes

Polar aprotic solvent with the highest known electrondonating power[1]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.0 °F - closed cup

Flash Point(C)

112.8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y. Ozari et al.
Journal of the Chemical Society. Chemical Communications, 295-295 (1974)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service