Skip to Content
Merck
All Photos(1)

Key Documents

93300

Sigma-Aldrich

Tris(2-ethylhexyl)amine

≥97.0% (GC)

Synonym(s):

2-Ethyl-N,N-bis(2-ethylhexyl)-1-hexanamine, TEHA, Tri(2-ethylhexyl)amine

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$403.00

$403.00


Available to ship on16 April 2025Details


Request a Bulk Order

Select a Size

Change View
250 ML
$403.00

About This Item

Linear Formula:
[CH3(CH2)3CH(C2H5)CH2]3N
CAS Number:
Molecular Weight:
353.67
Beilstein:
2087270
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$403.00


Available to ship on16 April 2025Details


Request a Bulk Order

Quality Level

Assay

≥97.0% (GC)

form

liquid

refractive index

n20/D 1.451

density

0.817 g/mL at 20 °C (lit.)

functional group

amine

SMILES string

CCCCC(CC)CN(CC(CC)CCCC)CC(CC)CCCC

InChI

1S/C24H51N/c1-7-13-16-22(10-4)19-25(20-23(11-5)17-14-8-2)21-24(12-6)18-15-9-3/h22-24H,7-21H2,1-6H3

InChI key

BZUDVELGTZDOIG-UHFFFAOYSA-N

Related Categories

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service