Skip to Content
Merck
All Photos(1)

Key Documents

931020

Sigma-Aldrich

9,9′-(4,4′-Sulfonylbis(4,1-phenylene))bis(3,6-dimethoxy-9H-carbazole)

≥97% (HPLC)

Synonym(s):

DMOC-DPS

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$485.00

$485.00


Estimated to ship on14 April 2025


Request a Bulk Order

Select a Size

Change View
250 MG
$485.00

About This Item

Empirical Formula (Hill Notation):
C40H32N2O6S
CAS Number:
Molecular Weight:
668.76
MDL number:
UNSPSC Code:
12352005
NACRES:
NA.23
Assay:
≥97% (HPLC)
grade:
sublimed grade
solubility:
chloroform: soluble
toluene: soluble

$485.00


Estimated to ship on14 April 2025


Request a Bulk Order

grade

sublimed grade

Quality Level

Assay

≥97% (HPLC)

band gap

0.21 eV (delta EST)

loss

0.5% TGA, > 250 °C (weight loss)

solubility

chloroform: soluble
toluene: soluble

fluorescence

λem 445 nm±10 nm in toluene

Orbital energy

HOMO 5.5 eV 
LUMO 2.45 eV 

λ

in toluene

UV absorption

λ: 370 nm±5 nm Amax

Application

It is commonly used as a hole transport material in organic electronic devices such as organic light-emitting diodes (OLEDs) and organic photovoltaic (OPV) devices. DMOC-DPS exhibits high hole mobility, possesses good thermal stability.
Use as OLED Materials: TADF Materials, Blue Dopant

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service