Skip to Content
Merck
All Photos(1)

Documents

930202

Sigma-Aldrich

3-(2,4,6-Trimethylbenzyl)-1-(2,6-diisopropylphenyl)-4,5-dimethylimidazolium chloride

≥95%

Synonym(s):

3-(2,6-diisopropylphenyl)-4,5-dimethyl-1-(2,4,6-trimethylbenzyl)-1H-imidazol-3-ium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H37ClN2
Molecular Weight:
425.05
UNSPSC Code:
12352005
NACRES:
NA.21

Quality Level

Assay

≥95%

form

powder

SMILES string

CC1=C(C)[N+](C2=C(C(C)C)C=CC=C2C(C)C)=CN1CC3=C(C)C=C(C)C=C3C.[Cl-]

Related Categories

Application

3-(2,4,6-Trimethylbenzyl)-1-(2,6-diisopropylphenyl)-4,5-dimethylimidazolium chloride is a highly hindered and flexible unsymmetrical NHC ligand for hydrofunctionalization reactions. When combined with a group 11 metal (Cu, Au, Ag), this ligand can be used in hydroelement additions, element-element additions, carbo-element additions, hydrations, hydroaminations, hydroarylations, hyroalkoxylations, hydrocarboxylations, cyclizations, hydrofluorinations, as well as a variety of combined processes using tandem reactions.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service