Skip to Content
Merck
All Photos(1)

Key Documents

929786

Sigma-Aldrich

PCL-PEG-PCL diacrylate

PCL average Mn 200, PEG average Mn 1000

Synonym(s):

Biodegradable triblock copolymer, PCL-b-PEG-b-PCL, PEG hydrogel, Polyethylene glycol, acrylate-PCL-PEG-PCL-acrylate

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$772.00

$772.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$772.00

About This Item

Linear Formula:
C2H3O2(C6H10O2)x(C2H4O)y(OC6H10O)zOC2H3
UNSPSC Code:
12162002
NACRES:
NA.25

$772.00


Check Cart for Availability

Request a Bulk Order

form

low-melting solid

Quality Level

mol wt

PCL average Mn 200
PEG average Mn 1000

contains

MEHQ as stabilizer

color

white to off-white

Ω-end

acrylate

α-end

acrylate

storage temp.

−20°C

SMILES string

O=C(OCCOC(CCCCCOC(C=C)=O)=O)CCCCCOC(C=C)=O

InChI

1S/C20H30O8/c1-3-17(21)25-13-9-5-7-11-19(23)27-15-16-28-20(24)12-8-6-10-14-26-18(22)4-2/h3-4H,1-2,5-16H2

InChI key

PCLZPCUCIICUMZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

PCL-PEG-PCL diacrylate, average Mn 100-1000-100, is a functionalized, amphiphilic triblock copolymer composed of a central hydrophilic, biocompatible poly(ethylene glycol) (PEG) block and two terminal hydrophobic, biodegradable poly(caprolactone) (PCL) blocks. PCL-PEG-PCL diacrylate can be crosslinked via free radical polymerization and/or photopolymerization to form a crosslinked hydrogel network. Hydrolysis of the esters of the PCL blocks permits degradation of the crosslinked networks.

Application

  • Tissue engineering
  • 3D printing
  • Drug delivery

Features and Benefits

  • PCL-PEG-PCL triblock copolymer
  • Hydrolytically degradable PCL blocks
  • Acrylate functional groups permit photopolymerization

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service