Skip to Content
Merck
All Photos(1)

Key Documents

929751

Sigma-Aldrich

Poly(ethylene glycol) diacrylate

average MN 3,400, acrylate, MEHQ as inhibitor

Synonym(s):

PEG bisacrylate, PEG diacrylate, PEG hydrogel, PEGDA, Polyethylene glycol

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$212.00

$212.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$212.00

About This Item

Linear Formula:
C3H3O(OCH2CH2)nC3H3O2
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.25

$212.00


Check Cart for Availability

Request a Bulk Order

Product Name

Poly(ethylene glycol) diacrylate, Mn 3,400, contains MEHQ as inhibitor

form

fine powder

Quality Level

mol wt

Mn 3,400

contains

MEHQ as inhibitor

color

white to off-white

solubility

deionized water: 20 wt. % at 23 °C

Ω-end

acrylate

α-end

acrylate

SMILES string

O(CCOC(=O)C=C)C(=O)C=C

InChI

1S/C8H10O4/c1-3-7(9)11-5-6-12-8(10)4-2/h3-4H,1-2,5-6H2

InChI key

KUDUQBURMYMBIJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(ethylene glycol) (PEG) diacrylate is a homobifunctional, hydrophilic, biocompatible polymer commonly used as a crosslinker in PEG hydrogels. Crosslinked networks can be formed via radical initiator induced polymerization or photopolymerization.

Application

PEG diacrylate has numerous applications including tissue engineering, 3D printing applications, surface modifications, PEGylation of proteins and peptides, and drug delivery.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service