929573
PCL-PEG-PCL diacrylate

average Mn 1000-10000-1000, Poly(caprolactone)-b-poly(ethylene glycol)-b-poly(caprolactone) diacrylate
Synonym(s):
Acrylate-PCL-PEG-PCL-acrylate, Biodegradable triblock copolymer, PEG hydrogel
Select a Size
About This Item
Quality Level
mol wt
PCL Mn 1000
PEG Mn 10000
12000 (theoretical)
average Mn 1000-10000-1000
contains
≤300 ppm MEHQ as inhibitor
color
white to off-white
Ω-end
acrylate
α-end
acrylate
storage temp.
−20°C
SMILES string
O=C(OCCOC(CCCCCOC(C=C)=O)=O)CCCCCOC(C=C)=O
InChI key
PCLZPCUCIICUMZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
- Tissue engineering
- 3D printing
- Drug delivery
Features and Benefits
- PCL-PEG-PCL triblock copolymer
- Hydrolytically degradable PCL blocks
- Acrylate functional groups permit photopolymerization
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service