Skip to Content
Merck
All Photos(1)

Key Documents

929549

Sigma-Aldrich

Ursodeoxycholic acid-2,2,4,4-d4

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$135.00
50 MG
$451.00

$135.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$135.00
50 MG
$451.00

About This Item

Empirical Formula (Hill Notation):
C24D4H36O4
CAS Number:
Molecular Weight:
396.60
MDL number:

$135.00


Check Cart for Availability

Request a Bulk Order

Quality Level

mass shift

M+4

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service