Skip to Content
Merck
All Photos(1)

Key Documents

927147

Sigma-Aldrich

BFMO

≥95%

Synonym(s):

N1,N2-Bis(furan-2-ylmethyl)oxalamide

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$110.00
1 G
$280.00

$110.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$110.00
1 G
$280.00

About This Item

Empirical Formula (Hill Notation):
C12H12N2O4
CAS Number:
Molecular Weight:
248.23
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.21

$110.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95%

form

powder

SMILES string

O=C(C(NCC1=CC=CO1)=O)NCC2=CC=CO2

InChI

1S/C12H12N2O4/c15-11(13-7-9-3-1-5-17-9)12(16)14-8-10-4-2-6-18-10/h1-6H,7-8H2,(H,13,15)(H,14,16)

InChI key

XRURWFXKCKASSN-UHFFFAOYSA-N

Application

BFMO is a ligand for the copper-catalyzed coupling of aryl halides and nitrogen heterocycles.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Copper(I) Oxide/N,N′-Bis[(2-furyl)methyl]oxalamide-Catalyzed Coupling of (Hetero)aryl Halides and Nitrogen Heterocycles at Low Catalytic Loading.
Pawar G G, et al.
Advanced Synthesis & Catalysis, 359(10), 1631-1636 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service