Skip to Content
Merck
All Photos(1)

Key Documents

926795

Sigma-Aldrich

3CzEPAIPN

≥95%

Synonym(s):

2,4,5-Tri(9H-carbazol-9-yl)-6-(ethyl(phenyl)amino)isophthalonitrile

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$254.00
500 MG
$883.00

$254.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$254.00
500 MG
$883.00

About This Item

Empirical Formula (Hill Notation):
C52H34N6
CAS Number:
Molecular Weight:
742.87
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.21

$254.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95%

form

powder

reaction suitability

reaction type: Photocatalysis
reagent type: catalyst

SMILES string

N#CC1=C(N2C(C=CC=C3)=C3C4=C2C=CC=C4)C(C#N)=C(N5C(C=CC=C6)=C6C7=C5C=CC=C7)C(N8C(C=CC=C9)=C9C%10=C8C=CC=C%10)=C1N(CC)C%11=CC=CC=C%11

InChI

1S/C52H34N6/c1-2-55(34-18-4-3-5-19-34)50-41(32-53)49(56-43-26-12-6-20-35(43)36-21-7-13-27-44(36)56)42(33-54)51(57-45-28-14-8-22-37(45)38-23-9-15-29-46(38)57)52(50)58-47-30-16-10-24-39(47)40-25-11-17-31-48(40)58/h3-31H,2H2,1H3

InChI key

KTBYFELTOZIXPZ-UHFFFAOYSA-N

Related Categories

Application

3CzEPAIPN is cyanoarene-based donor-acceptor photocatalyst.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jinhui Xu et al.
Journal of the American Chemical Society, 143(33), 13266-13273 (2021-08-26)
Since the seminal work of Zhang in 2016, donor-acceptor cyanoarene-based fluorophores, such as 1,2,3,5-tetrakis(carbazol-9-yl)-4,6-dicyanobenzene (4CzIPN), have been widely applied in photoredox catalysis and used as excellent metal-free alternatives to noble metal Ir- and Ru-based photocatalysts. However, all the reported photoredox

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service