Skip to Content
Merck
All Photos(1)

Key Documents

925802

Sigma-Aldrich

Silk fibroin

average mol wt 100 kDa (water soluble)

Synonym(s):

SilMA, Silk fibroin

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$194.00

$194.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$194.00

About This Item

UNSPSC Code:
12162002
NACRES:
NA.23

$194.00


Check Cart for Availability

Request a Bulk Order

mol wt

average mol wt 100 kDa (water soluble)

Quality Level

color

white to off-white

pH

6-8 (2% in water, when dissolved in water)

General description

Silk is a natural polymer composed of two proteins - fibroin and sericin.
Silk fibroin is used in biomedical applications due to its enhanced biocompatibility and biodegradability.

Silk fibroin (SF) demonstrates enhanced strength and stability compared to other common natural polymers due to its unique features including;
  • amphiphilicity
  • hydrophobicity
  • substantial hydrogen bonding
  • intrinsic low viscosity
  • high protein crystallinity by ß-sheet crystals.

Application

SF is a versatile material, that finds itself in multiple types of scaffolds, with extensive uses in inkjet, extrusion-based, laser-assisted, and DLP bioprinting tissue engineering applications.

Packaging

1 g in glass bottle

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Precisely printable and biocompatible silk fibroin bioink for digital light processing 3D printing.
Kim S H, et al.
Nature Communications, 9, 1620-1620 (2018)
Xiaohui Zhang et al.
Advanced drug delivery reviews, 61(12), 988-1006 (2009-08-01)
Electrospinning is a versatile technique that enables the development of nanofiber-based biomaterial scaffolds. Scaffolds can be generated that are useful for tissue engineering and regenerative medicine since they mimic the nanoscale properties of certain fibrous components of the native extracellular
Heesun Hong et al.
Biomaterials, 232, 119679-119679 (2019-12-23)
Three-dimensional printing with Digital Lighting Processing (DLP) printer has come into the new wave in the tissue engineering for regenerative medicine. Especially for the clinical application, it needs to develop of bio-ink with biocompatibility, biodegradability and printability. Therefore, we demonstrated
Soon Hee Kim et al.
Nature communications, 9(1), 1620-1620 (2018-04-26)
Although three-dimensional (3D) bioprinting technology has gained much attention in the field of tissue engineering, there are still several significant engineering challenges to overcome, including lack of bioink with biocompatibility and printability. Here, we show a bioink created from silk

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service