Skip to Content
Merck
All Photos(1)

Key Documents

925748

Sigma-Aldrich

N,N′-Bis(2-methylphenyl)-N,N′-bis(6-tert-butyldibenzofuran-4-yl)pyrene-3,8-diamine

≥97%

Synonym(s):

BD

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$767.00

$767.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$767.00

About This Item

Empirical Formula (Hill Notation):
C62H52N2O2
CAS Number:
Molecular Weight:
857.09
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.23

$767.00


Check Cart for Availability

Request a Bulk Order

description

PL: 453 nm

Quality Level

Assay

≥97%

form

powder or crystals

color

yellow

storage temp.

2-8°C

Application

N,N′-Bis(2-methylphenyl)-N,N′-bis(6-tert-butyldibenzofuran-4-yl)pyrene-3,8-diamine (BD) exhibits highly efficient blue emission with narrow FWHM. BD could be used as a blue fluorescent dopant in the fabrication of OLED device. The OLED device exhibits higher color purity (CIEx,y=(0.136, .0.100)) and excellent external quantum efficiency (EQE).
N,N′-Bis(2-methylphenyl)-N,N′-bis(6-tert-butyldibenzofuran-4-yl)pyrene-3,8-diamine can be used in the research areas as a component in organic field-effect transistors (OFETs) or organic photovoltaic cells, as an emitter or charge transport layer in organic light-emitting diodes (OLEDs), sensing applications, in optoelectronic devices such as lasers, photodetectors, or light-emitting electrochemical cells (LEECs) etc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aromatic amines having pyrene structures, organic electroluminescent materials containing them, and organic electroluminescent devices using them.
Mizuki, Y.; Ito, H.; et al
PCT Int. Appl. 94, 19338 (2013)
Stable pure-blue hyperfluorescence organic light-emitting diodes with high-efficiency and narrow emission
Chan, Chin-Yiu, et al.
Nature Photonics, 15, 203-207 (2021)
Junpeng Ji et al.
Nature communications, 12(1), 54-54 (2021-01-06)
Current power supply networks across the world are mostly based on three-phase electrical systems as an efficient and economical way for generation, transmission and distribution of electricity. Now, many electrically driven devices are relying on direct current or single-phase alternating

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service