Skip to Content
Merck
All Photos(1)

Key Documents

925144

Sigma-Aldrich

KB05yne

≥95%

Synonym(s):

4-(N-(4-Bromophenyl)acrylamido)-N-(hex-5-yn-1-yl)benzamide, Functionalized scout fragment

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$398.00

$398.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$398.00

About This Item

Empirical Formula (Hill Notation):
C22H21BrN2O2
Molecular Weight:
425.32
UNSPSC Code:
12352101
NACRES:
NA.22

$398.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95%

form

powder or solid

reaction suitability

reagent type: chemical modification reagent
reaction type: click chemistry

storage temp.

2-8°C

Application

KB05yne is a cysteine-reactive small-molecule fragment for chemoproteomic and ligandability studies for both traditionally druggable proteins as well as "undruggable," or difficult-to-target, proteins. This fragment electrophile is the functionalized version of KB02 (912131)

Related useful products may include: Cysteine-reactive fragments: KB02 (912131), KB03 (912654), KB05 (911798), sulfoxide (925136), CoLDR probe (923818) Functionalized scout fragments: KB02-COOH (925047), KB02yne (925225) Electrophilic degraders featuring scout fragments: KB02-SLF (914738), KB03-SLF (914975), KB05-SLF (913715), Biotin-SLF (914223) Cysteine-reactive probes for chemoproteomics: IA alkyne (924237), IA 5-TAMRA (925020), desthiobiotin iodoacetamide (923826), or biotin iodoacetamide (B2059)Related useful products may include: Cysteine-reactive fragments: KB02 (912131), KB03 (912654), KB05 (911798), sulfoxide (925136), CoLDR probe (923818) Functionalized scout fragments: KB02-COOH (925047), KB02yne (925225) Electrophilic degraders featuring scout fragments: KB02-SLF (914738), KB03-SLF (914975), KB05-SLF (913715), Biotin-SLF (914223) Cysteine-reactive probes for chemoproteomics: IA alkyne (924237), IA 5-TAMRA (925020), desthiobiotin iodoacetamide (923826), or biotin iodoacetamide (B2059)

Technology spotlight: Proteomic Ligandability Assessment

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrophilic PROTACs that degrade nuclear proteins by engaging DCAF16
Xiaoyu Zhang, et al
Nature Chemical Biology, 15(7), 737-746 (2019)
Vincent M Crowley et al.
ACS central science, 7(4), 613-623 (2021-06-01)
Covalent ligands are a versatile class of chemical probes and drugs that can target noncanonical sites on proteins and display differentiated pharmacodynamic properties. Chemical proteomic methods have been introduced that leverage electrophilic fragments to globally profile the covalent ligandability of
Xiaoyu Zhang et al.
Nature chemical biology, 15(7), 737-746 (2019-06-19)
Ligand-dependent protein degradation has emerged as a compelling strategy to pharmacologically control the protein content of cells. So far, however, only a limited number of E3 ligases have been found to support this process. Here, we use a chemical proteomic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service