Skip to Content
Merck
All Photos(1)

Key Documents

92494

Sigma-Aldrich

(+)-N,N-Diethylnorephedrine hydrochloride

≥95.0% (AT)

Synonym(s):

(1S,2R)-2-Diethylamino-1-phenyl-1-propanol hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C13H21NO · HCl
Molecular Weight:
243.77
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

Assay

≥95.0% (AT)

form

solid

storage temp.

2-8°C

SMILES string

Cl.CCN(CC)[C@H](C)[C@@H](O)c1ccccc1

InChI

1S/C13H21NO.ClH/c1-4-14(5-2)11(3)13(15)12-9-7-6-8-10-12;/h6-11,13,15H,4-5H2,1-3H3;1H/t11-,13-;/m1./s1

InChI key

KOCKBNBCNQXAJH-LOCPCMAASA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service