Skip to Content
Merck
All Photos(1)

Key Documents

924253

Sigma-Aldrich

Deoxazole-quat

≥95%

Synonym(s):

5,7-Di-tert-butyl-3-(4-(trifluoromethyl)phenyl)benzo[d]oxazol-3-ium tetrafluoroborate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$297.00
5 G
$911.00

$297.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$297.00
5 G
$911.00

About This Item

Empirical Formula (Hill Notation):
C22H25BF7NO
CAS Number:
Molecular Weight:
463.24
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.21

$297.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95%

form

powder

mp

230-234 °C

SMILES string

CC(C)(C)C1=CC([N+](C2=CC=C(C(F)(F)F)C=C2)=CO3)=C3C(C(C)(C)C)=C1.F[B-2](F)(F)F

Application

Deoxazole-quat is an N-heterocyclic carbene salt which activates tertiary free alcohols for a C-C cross coupling reaction with aryl halides. The MacMillan group developed a mild, robust, and selective metallaphotoredox-based cross-coupling platform for the deoxygenative coupling for free alcohols using Deoxazole-quat, a Ni catalyst, and [Ir(dtbbpy)(ppy)2]PF6.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhe Dong et al.
Nature, 598(7881), 451-456 (2021-09-01)
Metal-catalysed cross-couplings are a mainstay of organic synthesis and are widely used for the formation of C-C bonds, particularly in the production of unsaturated scaffolds1. However, alkyl cross-couplings using native sp3-hybridized functional groups such as alcohols remain relatively underdeveloped2. In particular

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service