Skip to Content
Merck
All Photos(1)

Key Documents

924075

Sigma-Aldrich

NHS ester-a-TCO

Synonym(s):

(E)-N-Hydroxysuccinyl 2-(1-ax-hydroxycyclooct-4-en-1-yl)acetate, 2,5-Dioxopyrrolidin-1-yl (R,E)-2-(1-hydroxycyclooct-4-en-1-yl)acetate, Amine-reactive axial 5-hydroxy-trans-cyclooctene

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$353.00
50 MG
$873.00

$353.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$353.00
50 MG
$873.00

About This Item

Empirical Formula (Hill Notation):
C14H19NO5
Molecular Weight:
281.30
UNSPSC Code:
12352108
NACRES:
NA.22

$353.00


Check Cart for Availability

Request a Bulk Order

reaction suitability

reaction type: click chemistry
reactivity: amine reactive
reagent type: chemical modification reagent
reagent type: cross-linking reagent

Quality Level

functional group

NHS ester

storage temp.

−20°C

SMILES string

O[C@@]1(CC(ON2C(CCC2=O)=O)=O)CC/C=C/CCC1

InChI key

HKYXURAVXKJCHH-AWEZNQCLSA-N

Application

NHS ester-a-TCO is an axial trans-cyclooctene (a-TCO) for bioconjugation and protein labeling. NHS ester-a-TCO has an amine-reactive NHS ester, and the a-TCO reacts rapidly with tetrazines through Diels–Alder reactions, a type of bioorthogonal, copper-free click chemistry. As demonstrated by the Joe Fox lab in Pigga, et al, a-TCOs are more reactive and hydrophilic than standard TCOs, providing favorable physiochemical properties for live-cell and in vivo labeling.

Other new a-TCOs:
  • TAMRA-a-TCO (924083)
  • Alkyne-a-TCO (924091)
  • Amine-a-TCO (924113)

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jessica E Pigga et al.
Angewandte Chemie (International ed. in English), 60(27), 14975-14980 (2021-03-21)
trans-Cyclooctenes (TCOs) are essential partners in the fastest known bioorthogonal reactions, but current synthetic methods are limited by poor diastereoselectivity. Especially hard to access are hydrophilic TCOs with favorable physicochemical properties for live cell or in vivo experiments. Described is a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service