Skip to Content
Merck
All Photos(1)

Key Documents

922277

Sigma-Aldrich

TpOx-B15C5

Synonym(s):

TpOx-Crown Ether Series

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$533.00

$533.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$533.00

About This Item

Empirical Formula (Hill Notation):
C58H79NO11
CAS Number:
Molecular Weight:
966.25
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.23

$533.00


Check Cart for Availability

Request a Bulk Order

description

Polymorph crystalline phase: Solid, Discotic Liquid Crystal, Isotropic

Quality Level

solubility

soluble (DCM,THF > 1 mg /mL, MeCN, DMSO < 1 mg /mL)

λmax

270 nm in ethyl acetate

εmax

86,000 M-1cm-1 in ethyl acetate

fluorescence

λem 457 nm, quantum yield 0.43 (pSS(nm) = 187; Fluorescence lifetime (ns) = 4.23)

storage temp.

2-8°C

General description

TpOx-B15C5 is a UV excitable fluorescent material with a polycyclic aromatic donor-acceptor structure where the triphenoxazole core acts as a donor and the aromatic group on the two position of the oxazole (Benzo-15-crown-5) act as the acceptor group. The push-pull, donor-acceptor, structure facilitates intramolecular charge transfer in the excited state that results in a 187 nm emission Stokes Shift. TpOx-B15C5 is designed for 355 nm excitation with emission at 457 nm with quantum yield of 0.43 and undergo fluorescent modulation upon binding metal cations in an appended crown ether moiety. TpOx-B15C5 has potential uses in metal cation sensing, fluorescent dye staining, organic electronic and photonics, and imaging applications.

Application

Fluorescent materials have a range of unique properties that allows for potential use in a range of applications:

  • Metal cation sensing
TpOx-B15C5 undergo fluorescent modulation upon binding metal cations in an appended crown ether moiety. TpOxB15C5 shows binding with Li+, K+ and Mg2+.

  • Fluorescent dye staining
UV fluorescent materials can be used to dope polymer and resin composites, including 3D applications, and further identified using fluorescence emission.

  • Organic semiconductor for organic electronic and photonic applications
These organic discotic liquid crystals display columnar hexagonal mesophase, and were incorporated into OPV device as a hole transport layer and donor material. Similar compounds have been used in organic light emitting devices (OLEDs), organic photovoltaic device (OPVs) and organic field-effect transistor (OFETs) as hole transport material.

  • Multi-photon microscopy
These UV fluorescent materials have been used in multi-photon microscopy, showing remarkable image resolution relative to commercial fluorophores in the multiphoton microscopy imaging. By perfusing it into an ex-planted human liver, it was possible to observe fluorophore staining of the hepatocytes and T-cells in the liver vasculature. During the live liver imaging these probes have been shown to perfuse effectively in human tissue and emit brightly without significant cytotoxicity during the course of the experiment.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Triphenoxazoles exhibit tunable properties, liquid crystallinity, and potential for modification.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service